Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPT1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519021
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519021 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519021 Supplier Invitrogen™ Supplier No. PA519021
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPT1C
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CPT1C
Gene Accession No. Q8TCG5
Gene Alias 6530437J22Rik; 9630004I06Rik; Carnitine O-palmitoyltransferase 1, brain isoform; carnitine O-palmitoyltransferase I, brain isoform; carnitine palmitoyltransferase 1, brain; carnitine palmitoyltransferase 1c; carnitine palmitoyltransferase I related C; CATL1; CPT IC; CPT I-C; CPT1-B; Cpt1c; CPT1P; CPTI-B; CPTIC; FLJ23809; SPG73
Gene Symbols CPT1C
Host Species Goat
Immunogen Synthetic peptide sequence (KSSTKTDSHRLGQH) corresponding to the internal amino acids of CPT1C (aa 737-751).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126129
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.