Learn More
Invitrogen™ CPT1C Polyclonal Antibody

Description
CPT1C Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | CPT1C |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.27 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CPT1C |
| Gene Accession No. | Q8TCG5 |
| Gene Alias | 6530437J22Rik; 9630004I06Rik; Carnitine O-palmitoyltransferase 1, brain isoform; carnitine O-palmitoyltransferase I, brain isoform; carnitine palmitoyltransferase 1, brain; carnitine palmitoyltransferase 1c; carnitine palmitoyltransferase I related C; CATL1; CPT IC; CPT I-C; CPT1-B; Cpt1c; CPT1P; CPTI-B; CPTIC; FLJ23809; SPG73 |
| Gene Symbols | CPT1C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.