Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPT1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589482 Supplier Invitrogen™ Supplier No. PA589482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYRHRLNRQE IPPTLLMGMR PLCSAQYEKI FNTTRIPGVQ KDYIRHLHDS QHVAVFHRGR FFRMGTHSRN SLLSPRALEQ QFQRILDDPS PACPHEEHLA ALTAAPRGTW AQVRTSLKTQ AAEALEAVEG A; Positive Samples: Mouse brain, Mouse testis, Mouse kidney, Rat brain, Rat testis, Rat kidney; Cellular Location: Cell junction, Cell projection, Endoplasmic reticulum, Mitochondrion outer membrane, Multi-pass membrane protein, axon, dendrite, synapse.

This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPT1C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 7.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CPT1C
Gene Accession No. F1LN46, Q8BGD5, Q8TCG5
Gene Alias 6530437J22Rik; 9630004I06Rik; Carnitine O-palmitoyltransferase 1, brain isoform; carnitine O-palmitoyltransferase I, brain isoform; carnitine palmitoyltransferase 1, brain; carnitine palmitoyltransferase 1c; carnitine palmitoyltransferase I related C; CATL1; CPT IC; CPT I-C; CPT1-B; Cpt1c; CPT1P; CPTI-B; CPTIC; FLJ23809; SPG73
Gene Symbols CPT1C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 270-400 of human CPT1C (NP_0011295241).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126129, 308579, 78070
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.