Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPT2 Recombinant Rabbit Monoclonal Antibody (9V6M9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535056
Description
Immunogen sequence: LRHLAAAKGI ILPELYLDPA YGQINHNVLS TSTLSSPAVN LGGFAPVVSD GFGVGYAVHD NWIGCNVSSY PGRNAREFLQ CVEKALEDMF DALEGKSIKS.
Carnitine palmitoyltransferase II precursor (CPT2) is a nuclear protein which is transported to the mitochondrial inner membrane. CPT2 together with carnitine palmitoyltransferase I oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders.
Specifications
CPT2 | |
Recombinant Monoclonal | |
0.28 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18886, P23786, P52825 | |
CPT2 | |
A synthetic peptide corresponding to a sequence within amino acids 559-658 of human CPT2 (P23786). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
9V6M9 | |
Unconjugated | |
CPT2 | |
AI323697; Carnitine O-palmitoyltransferase 2, mitochondrial; carnitine palmitoyltransferase 2; carnitine palmitoyltransferase II; CPT II; CPT1; CPT2; Cpt-2; CPTASE; CPTII; IIAE4; OTTHUMP00000010493; testicular secretory protein Li 13 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12896, 1376, 25413 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction