Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110953
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110953 100 μL
PIPA5110952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5110953 Supplier Invitrogen™ Supplier No. PA5110953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMVECSKYHG QLTKEAAMGQ GFDRHLFALR HLAAAKGIIL PELYLDPAYG QINHNVLSTS TLSSPAVNLG GFAPVVSDGF GVGYAVHDNW IGCNVSSYP Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 83%.

Carnitine palmitoyltransferase II precursor (CPT2) is a nuclear protein which is transported to the mitochondrial inner membrane. CPT2 together with carnitine palmitoyltransferase I oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPT2
Gene Accession No. P23786
Gene Alias AI323697; Carnitine O-palmitoyltransferase 2, mitochondrial; carnitine palmitoyltransferase 2; carnitine palmitoyltransferase II; CPT II; CPT1; CPT2; Cpt-2; CPTASE; CPTII; IIAE4; OTTHUMP00000010493; testicular secretory protein Li 13
Gene Symbols CPT2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CPT2. Recombinant protein control fragment (Product #RP-94317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1376
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.