Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPT2 Rabbit anti-Human, Mouse, Rat, Clone: 9V6M9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | CPT2 |
---|---|
Clone | 9V6M9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CPT2 Monoclonal antibody specifically detects CPT2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CPT2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1376 | |
IgG | |
Affinity purified |
9V6M9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
carnitine O-palmitoyltransferase 2, mitochondrial, carnitine palmitoyltransferase 2, Carnitine palmitoyltransferase IIEC 2.3.1.21, CPT II, CPT1, CPTASE | |
A synthetic peptide corresponding to a sequence within amino acids 559-658 of human CPT2 (P23786). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title