Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPVL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588502 Supplier Invitrogen™ Supplier No. PA588502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YVVTSNMTLR DRDFPWTTTL SMLYIDNPVG TGFSFTDDTH GYAVNEDDVA RDLYSALIQF FQIFPEYKNN DFYVTGESYA GKYVPAIAHL IHSLNPVREV KINLNGIAIG DGYSDPESII GGYAEFLYQI GLLDEKQKKY FQKQCHECIE HIRKQNWFEA FEILDKLLDG DLTSDPSYFQ NVTGCSNYYN FLRCTEPEDQ LYYVKFLSLP EVRQAIHVGN QTFNDGTIVE KYLREDTVQS VKPWLTEIMN NYKVLIYNGQ LDIIVAAALT ERSLMGMDWK GSQEYKKAEK KVWKIFKSDS EVAGYIRQAG DFHQVIIRGG GHILPYDQPL RAFDMINRFI YGKGWDPYVG; Positive Samples: Mouse brain, Rat testis, Rat brain.

The protein encoded by this gene is a carboxypeptidase and bears strong sequence similarity to serine carboxypeptidases. Carboxypeptidases are a large class of proteases that act to cleave a single amino acid from the carboxy termini of proteins or peptides. The exact function of this protein, however, has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPVL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Cpvl
Gene Accession No. Q4QR71, Q9D3S9
Gene Alias 4933436L16Rik; carboxypeptidase WUG; carboxypeptidase, vitellogenic like; carboxypeptidase, vitellogenic-like; CP-Mac carboxypeptidase; CPVL; HVLP; probable serine carboxypeptidase CPVL; PSEC0124; tcag7.235; UNQ197/PRO223; VCP-like protein; vitellogenic carboxypeptidase-like protein; VLP
Gene Symbols Cpvl
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 127-476 of human CPVL (NP_0619022).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 502774, 71287
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.