Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPVL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561079
Description
Immunogen sequence: MNNYKVLIYN GQLDIIVAAA LTERSLMGMD WKGSQEYKKA EKKVWKIFKS DSEVAGYIRQ AGDFHQVIIR GGGHILPYDQ PLRAFDMINR FIYGKGWDPY VG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
The protein encoded by this gene is a carboxypeptidase and bears strong sequence similarity to serine carboxypeptidases. Carboxypeptidases are a large class of proteases that act to cleave a single amino acid from the carboxy termini of proteins or peptides. The exact function of this protein, however, has not been determined.
Specifications
CPVL | |
Polyclonal | |
Unconjugated | |
Cpvl | |
4933436L16Rik; carboxypeptidase WUG; carboxypeptidase, vitellogenic like; carboxypeptidase, vitellogenic-like; CP-Mac carboxypeptidase; CPVL; HVLP; probable serine carboxypeptidase CPVL; PSEC0124; tcag7.235; UNQ197/PRO223; VCP-like protein; vitellogenic carboxypeptidase-like protein; VLP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54504 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H3G5 | |
Cpvl | |
Recombinant protein corresponding to Human CPVL. Recombinant protein control fragment (Product #RP-102984). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction