Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPXM Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988480
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8480 Supplier Bioss Supplier No. BS8341R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the M14 family of zinc carboxypeptidases, however, the protein has no detectable carboxypeptidase activity. The encoded protein is thought to be an extracellular and/or membrane protein, and may be involved in cell-cell interactions.

Specifications

Antigen CPXM
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CPXM1
Gene Accession No. Q96SM3
Gene Alias AA986902; carboxypeptidase X (M14 family); carboxypeptidase X (M14 family), member 1; carboxypeptidase X 1 (M14 family); carboxypeptidase-like protein X1; CPX1; Cpx-1; CPXM; CPXM1; metallocarboxypeptidase 1; Metallocarboxypeptidase CPX-1; Probable carboxypeptidase X1; UNQ3015/PRO9782
Gene Symbols CPXM1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CPXM.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56265
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.