Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPXM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548021
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548021 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548021 Supplier Invitrogen™ Supplier No. PA548021
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse CPXM is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the M14 family of zinc carboxypeptidases, however, the protein has no detectable carboxypeptidase activity. The encoded protein is thought to be an extracellular and/or membrane protein, and may be involved in cell-cell interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPXM1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CPXM1
Gene Accession No. Q96SM3
Gene Alias AA986902; carboxypeptidase X (M14 family); carboxypeptidase X (M14 family), member 1; carboxypeptidase X 1 (M14 family); carboxypeptidase-like protein X1; Cpx1; Cpx-1; Cpxm; Cpxm1; metallocarboxypeptidase 1; metallocarboxypeptidase CPX-1; Probable carboxypeptidase X1; UNQ3015/PRO9782
Gene Symbols CPXM1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase X1/CPXM1 Ala21-Asp734.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56265
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.