Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPXM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531412 Supplier Invitrogen™ Supplier No. PA531412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (90%), Rat (92%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CPXM2 may be involved in cell-cell interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPXM2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CPXM2
Gene Accession No. Q8N436
Gene Alias 4632435C11Rik; carboxypeptidase Hlo; carboxypeptidase X (M14 family), member 2; carboxypeptidase X 2 (M14 family); carboxypeptidase X2; carboxypeptidase-like protein X2; Cpx2; CPX-2; CPXM2; cytosolic carboxypeptidase; inactive carboxypeptidase-like protein X2; metallocarboxypeptidase 2; RGD1310955; UNQ676; UNQ676/PRO1310
Gene Symbols CPXM2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human CPXM2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.