Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPXM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575876 Supplier Invitrogen™ Supplier No. PA575876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

CPXM2 may be involved in cell-cell interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPXM2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.3
Gene CPXM2
Gene Accession No. Q8N436, Q9D2L5
Gene Alias 4632435C11Rik; carboxypeptidase Hlo; carboxypeptidase X (M14 family), member 2; carboxypeptidase X 2 (M14 family); carboxypeptidase X2; carboxypeptidase-like protein X2; Cpx2; CPX-2; CPXM2; cytosolic carboxypeptidase; inactive carboxypeptidase-like protein X2; metallocarboxypeptidase 2; RGD1310955; UNQ676; UNQ676/PRO1310
Gene Symbols CPXM2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human CPXM2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119587, 293566, 55987
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.