Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPXM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103709
Description
Antibody detects endogenous levels of total CPXM2.
CPXM2 may be involved in cell-cell interactions.
Specifications
| CPXM2 | |
| Polyclonal | |
| Unconjugated | |
| CPXM2 | |
| 4632435C11Rik; carboxypeptidase Hlo; carboxypeptidase X (M14 family), member 2; carboxypeptidase X 2 (M14 family); carboxypeptidase X2; carboxypeptidase-like protein X2; Cpx2; CPX-2; CPXM2; cytosolic carboxypeptidase; inactive carboxypeptidase-like protein X2; metallocarboxypeptidase 2; RGD1310955; UNQ676; UNQ676/PRO1310 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 119587, 55987 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N436, Q9D2L5 | |
| CPXM2 | |
| A synthesized peptide derived from human CPXM2(Accession Q8N436), corresponding to amino acid residues N337-E387. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction