Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CPYPP
GREENER_CHOICESDP

Catalog No. 456850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
456850 50 mg
456810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 456850 Supplier Tocris Bioscience™ Supplier No. 4568/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

DOCK2-Rac1 interaction inhibitor

CPYPP is a DOCK2-Rac1 interaction inhibitor; suppresses DOCK2-induced Rac1 activation. Also inhibits DOCK1 and DOCK5. Exhibits selectivity over DOCK9. Reduces chemotactic responses in vitro and in vivo, and activates T cells in vitro. Also attenuates HER2-mediated breast cancer cell migration in vitro.

Specifications

Chemical Name or Material Rho GTPase Inhibitors
CAS 310460-39-0
Quantity 50 mg
Target Small G Protein Inhibitors
Molecular Formula C18H13ClN2O2
Formula Weight 324.76
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.