Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568134 Supplier Invitrogen™ Supplier No. PA568134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Rat samples.

CPZ is a member of the metallocarboxypeptidase family. This enzyme displays carboxypeptidase activity towards substrates with basic C-terminal residues. It is most active at neutral pH and is inhibited by active site-directed inhibitors of metallocarboxypeptidases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPZ
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CPZ
Gene Accession No. O54858, Q66K79
Gene Alias AV136066; carboxypeptidase Z; Cpz; metallocarboxypeptidase Z
Gene Symbols CPZ
Host Species Rabbit
Immunogen Synthesized peptide derived from human CPZ.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83575, 8532
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.