Learn More
CR1 Mouse anti-Human, Clone: CR1/802, Abnova™
Mouse monoclonal antibody raised against full length recombinant human CR1.
Supplier: Abnova Corporation MAB13262
Description
This gene is a member of the receptors of complement activation (RCA) family and is located in the 'cluster RCA' region of chromosome 1. The gene encodes a monomeric single-pass type I membrane glycoprotein found on erythrocytes, leukocytes, glomerular podocytes, and splenic follicular dendritic cells. The Knops blood group system is a system of antigens located on this protein. The protein mediates cellular binding to particles and immune complexes that have activated complement. Decreases in expression of this protein and/or mutations in its gene have been associated with gallbladder carcinomas, mesangiocapillary glomerulonephritis, systemic lupus erythematosus and sarcoidosis. Mutations in this gene have also been associated with a reduction in Plasmodium falciparum rosetting, conferring protection against severe malaria. Alternate allele-specific splice variants, encoding different isoforms, have been characterized. Additional allele specific isoforms, including a secreted form, have been described but have not been fully characterized. [provided by RefSeq]
Specifications
CR1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
C3BR/CD35/KN | |
Mouse | |
Protein A/G purification | |
RUO | |
1378 | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
CR1/802 | |
Mouse monoclonal antibody raised against full length recombinant human CR1. | |
In 10mM PBS. | |
CR1 | |
Recombinant protein corresponding to full length human CR1. | |
100 μg | |
Primary | |
This monoclonal antibody does not block CR1 activity. It is highly specific to CR1 and shows no cross-reaction with CR2. | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.