Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CR16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559481
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559481 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559481 Supplier Invitrogen™ Supplier No. PA559481
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELSSKSQQAT AWTPTQQPGG QLRNGSLHII DDFESKFTFH SVEDFPPPDE YKPCQKIYPS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.

May be a regulator of cytoskeletal organization. May have a role in spermatogenesis. Target protein binds SH3 domains; may play a role in MAP kinase mediated signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CR16
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WIPF3
Gene Accession No. A6NGB9
Gene Alias corticosteroids and regional expression protein 16; Corticosteroids and regional expression protein 16 homolog; CR16; FLJ36931; SH3 domain binding protein CR16; WAS/WASL interacting protein family member 3; WAS/WASL interacting protein family, member 3; WAS/WASL-interacting protein family member 3; Wipf3
Gene Symbols WIPF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CR16. Recombinant protein control fragment (Product #RP-98475).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644150
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.