Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CR16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559449
Description
Immunogen sequence: QIESSKGTNK EGGGSANTRG ASTPPTLGDL FAGGFPVLRP AGQRDVAGGK TGQGPGSRAP SPRLPNKTIS GPLIPPASPR LGNTS Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 85%.
May be a regulator of cytoskeletal organization. May have a role in spermatogenesis. Target protein binds SH3 domains; may play a role in MAP kinase mediated signal transduction.
Specifications
CR16 | |
Polyclonal | |
Unconjugated | |
WIPF3 | |
corticosteroids and regional expression protein 16; Corticosteroids and regional expression protein 16 homolog; CR16; FLJ36931; SH3 domain binding protein CR16; WAS/WASL interacting protein family member 3; WAS/WASL interacting protein family, member 3; WAS/WASL-interacting protein family member 3; Wipf3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
644150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NGB9 | |
WIPF3 | |
Recombinant protein corresponding to Human CR16. Recombinant protein control fragment (Product #RP-98476). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction