Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CR16 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572271
Description
May be a regulator of cytoskeletal organization. May have a role in spermatogenesis (By similarity).
Specifications
CR16 | |
Polyclonal | |
Unconjugated | |
WIPF3 | |
WAS/WASL-interacting protein family member 3; Corticosteroids and regional expression protein 16 homolog; WIPF3; CR16 | |
Rabbit | |
Protein A | |
RUO | |
259242, 644150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.65 mg/mL | |
PBS with 0.09% sodium azide | |
A6NGB9, Q9Z0G8 | |
WIPF3 | |
KLH conjugated synthetic peptide between 109-142 amino acids from the N-terminal region of human WIPF3 | |
200 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction