Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cr2, anti-Mouse, Clone: 8C12, Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00124
Specifications
| Cr2 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.02% Proclin 300). | |
| C3DR/CD21/CD35/Cr-1/Cr-2/Cr1/MGC130295 | |
| Original antibody is raised against native purified mouse Cr2 protein. | |
| RUO | |
| 12902 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 8C12 | |
| Flow Cytometry (1 ug/mL) Immunohistochemistry (Frozen sections) (5 ug/mL) Immunoprecipitation (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| complement receptor 2 | |
| Cr2 | |
| 200 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction