Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CR2 Mouse anti-Human, Unconjugated, Clone: CR2/1952, Abnova™

Catalog No. 89337653 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-653 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-653 Supplier Abnova Corporation Supplier No. MAB21160
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CR2 Monoclonal antibody specifically detects CR2 in Human samples. It is validated for IHC-P.

Specifications

Antigen CR2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CR2/1952
Conjugate Unconjugated
Description complement component (3d/Epstein Barr virus) receptor 2
Gene CR2
Gene Alias C3DR, CD21, SLEB9
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 44-196 of human CR2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1380
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.