Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CR2 Rabbit anti-Human, Unconjugated, Clone: RM372, Abnova™

Catalog No. 89339221 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-221 Supplier Abnova Corporation Supplier No. MAB22004
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CR2 Monoclonal antibody specifically detects CR2 in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen CR2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM372
Conjugate Unconjugated
Description complement component (3d/Epstein Barr virus) receptor 2
Gene CR2
Gene Alias C3DR, CD21, SLEB9
Host Species Rabbit
Immunogen A peptide corresponding to C-terminus of human CR2
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 1380
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.