Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRABP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597616
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597616 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597616 Supplier Invitrogen™ Supplier No. PA597616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRABP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRABP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CRABP1
Gene Accession No. P29762, P62965
Gene Alias AI326249; cCRABP-I; cellular retinoic acid binding protein 1; cellular retinoic acid binding protein I; cellular retinoic acid binding protein I (CRABP-I) homologue; cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; cellular retinoic-acid-binding protein; CRABP; Crabp1; Crabp-1; CrabpI; CRABP-I; RBP5; Rbp-5; retinoic acid-binding protein
Gene Symbols CRABP1
Host Species Rabbit
Immunogen Recombinant Human Cellular retinoic acid-binding protein 1 protein (2-137AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12903, 1381
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.