Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRABP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103724 Supplier Invitrogen™ Supplier No. PA5103724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CRABP1.

This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRABP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CRABP1
Gene Accession No. P29762, P62965, P62966
Gene Alias AI326249; cCRABP-I; cellular retinoic acid binding protein 1; cellular retinoic acid binding protein I; cellular retinoic acid binding protein I (CRABP-I) homologue; cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; cellular retinoic-acid-binding protein; CRABP; Crabp1; Crabp-1; CrabpI; CRABP-I; RBP5; Rbp-5; retinoic acid-binding protein
Gene Symbols CRABP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CRABP1(Accession P29762), corresponding to amino acid residues Q50-L100.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12903, 1381, 25061
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.