Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRABP2 Antibody (101), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290627PCP
Description
CRABP2 Monoclonal antibody specifically detects CRABP2 in Mouse samples. It is validated for ELISASpecifications
CRABP2 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
101 | |
ELISA | |
cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
Rabbit immunized with purified, recombinant Mouse CRABP2 (Uniprot#: P22935, Pro 2-Glu 138) | |
0.1 mL | |
Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
1382 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction