Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRABP2 Antibody (OTI10D6), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270448AF594
Description
CRABP2 Monoclonal specifically detects CRABP2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
CRABP2 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CRABP2 | |
Full length recombinant protein of human CRABP2 (NP_001869) produced in E.coli. | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI10D6 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
Mouse | |
0.1 mL | |
1382 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction