Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRABP2 Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA551146
Description
Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.
Specifications
CRABP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
P29373, P51673 | |
CRABP2 | |
Recombinant Human CRABP2 protein, Met1-Glu138 (Accession #P29373). | |
20 μL | |
Primary | |
Human, Rat, Pig | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
C1 | |
Unconjugated | |
CRABP2 | |
AI893628; cellular lipid-binding protein; cellular retinoic acid binding protein 2; cellular retinoic acid binding protein II; cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; Crabp2; Crabp-2; CrabpII; CRABP-II; RBP6; RP11-66D17.4 | |
Mouse | |
Protein A/G | |
RUO | |
100155151, 1382, 29563 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction