Learn More
CRABP2 Mouse anti-Human, Clone: OTI10D6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800069

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
A number of specific carrier proteins for members of the vitamin A family have been discovered. Cellular retinoic acid binding proteins (CRABP) are low molecular weight proteins whose precise function remains unknown. The inducibility of the CRABP2 gene suggests that this isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. Recognizes mouse and human CRABP2. No crossreactivity with CRABP1.Specifications
CRABP2 | |
Monoclonal | |
Unconjugated | |
CRABP2 | |
CRABP-II, RBP6 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1382 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI10D6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P29373 | |
CRABP2 | |
Full length human recombit protein of human CRABP2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.