Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CRACC, Rat, Unlabeled, Clone: 4G2,

Catalog No. MABF917MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF917MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF917MI Supplier MilliporeSigma™ Supplier No. MABF917
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SLAM family member 7 (UniProt Q8BHK6; also known as CD2-like receptor-activating cytotoxic cells, CRACC, Leukocyte cell-surface antigen, Novel Ly9, CD319) is encoded by the Slamf7 (also known as Cs1) gene (Gene ID 75345) in murine species. CRACC/CS1 belongs to the 6-member (SLAM/CD150, Ly-9/CD229, 2B4/CD244, CD84, NTB-A/CD352, CRACC) signaling lymphocytic activation molecule family (SLAMF) of receptors broadly expressed in hematopoietic cells. 2B4 and CD48 interact with each other, while rest of the family members act as self-ligands, where two identical molecules on the surface of two different cells can interact as receptor and ligand to each other. CRACC is synthesized with a signal peptide sequence (a.a. 1-22), the removal of which yields the mature CRACC with an extracellular region (a.a. 23-224) that contains a V-type and a C2-type Ig-like domain (a.a. 27-112 and 128-203), followed by a single transmembrane domain (a.a. 225-245) and a cytoplasmic tail (a.a. 246-333). CRACC cytoplasmic domain contains one immunoreceptor tyrosine-based switch motif (ITSM; 278-TFYSTV-283, numbering based on mature murine sequence), which upon phosphorylation (pY300) recruits SAP family adaptor EWS-Fli1-activated transcript-2 (EAT-2) for signaling transduction. CRACC can also be phosphorylated on Tyr280. Unlike pY300, pY280 mediates the inhibitory function of CRACC in NK cells. In addition to NK cells, evidence indicates that CRACC is also involved in regulating B- and T-cell functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRACC
Applications Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone 4G2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1κ antibody in PBS without preservatives.
Gene Slamf7, Cs1
Gene Accession No. Q8BHK6
Host Species Rat
Immunogen Human IgG1 Fc -tagged recombinant mouse CRACC extracellular domain.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_653122.2
Test Specificity Clone 4G2 reacted with murine T lymphoma BI-141 transfectants expressing mouse CRACC but not with those transfected with other members of the SLAM family (CD84, 2B4, Ly9, Ly108 or SLAM) (Cruz-Munoz, M.E., et al. (2009). Nat. Immunol. 10(3):297-305). Expected to react with all four spliced isoforms of murine CRACC reported by UniProt (Q8BHK6).
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.