Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRACC/SLAMF7 Antibody (002), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289961PE
Description
CRACC/SLAMF7 Monoclonal antibody specifically detects CRACC/SLAMF7 in Human samples. It is validated for ELISASpecifications
CRACC/SLAMF7 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
002 | |
ELISA | |
19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7 | |
Rabbit immunized with purified, recombinant Human CRACC/SLAMF7 (Accession#: NP_067004.3, Met1-Met226) | |
0.1 mL | |
Immunology | |
57823 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction