Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRACC/SLAMF7 Antibody (007), DyLight 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290478G
Description
CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Mouse samples. It is validated for ELISA.Specifications
CRACC/SLAMF7 | |
Monoclonal | |
DyLight 488 | |
19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CRACC/SLAMF7 extracellular domain (NP_653122.2; Met 1-Gly 224). | |
0.1 mL | |
Immunology | |
57823 | |
Mouse | |
Purified |
ELISA | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human SLAMF7/ CRACC | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction