Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRACC/SLAMF7 Antibody (235614), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1906MFV500
Description
CRACC/SLAMF7 Monoclonal antibody specifically detects CRACC/SLAMF7 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| CRACC/SLAMF7 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 57823 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 235614 | |
| Flow Cytometry, CyTOF-ready | |
| 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7 | |
| Mouse myeloma cell line NS0-derived recombinant human CRACC/SLAMF7, Lys27-Ser225, Accession # Q9NQ25 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction