Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRACC/SLAMF7 Antibody (OTI1F1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272107
Description
CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CRACC/SLAMF7 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Flow Cytometry, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin | |
19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7 | |
Mouse | |
37.2 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1F1 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
SLAMF7 | |
Full length human recombinant protein of human SLAMF7 (NP_067004) produced in HEK293T cell. | |
Affinity Purified | |
Immunology | |
57823 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction