Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRALBP Antibody (B2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10074392
Description
CRALBP Monoclonal specifically detects CRALBP in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CRALBP | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 0.02 ug/ml, Simple Western, Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1 ug/ml | |
Cellular retinaldehyde-binding protein, cellular retinaldehyde-binding protein-1, CRALBPretinaldehyde-binding protein 1, MGC3663, retinaldehyde binding protein 1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Primate | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
B2 | |
Unconjugated | |
P12271 | |
RLBP1 | |
Human recombinant CRALBP. | |
100 μg | |
Vision | |
6017 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction