Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRALBP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | CRALBP |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CRALBP Polyclonal specifically detects CRALBP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CRALBP | |
Polyclonal | |
Rabbit | |
Vision | |
Cellular retinaldehyde-binding protein, cellular retinaldehyde-binding protein-1, CRALBPretinaldehyde-binding protein 1, MGC3663, retinaldehyde binding protein 1 | |
RLBP1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
6017 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PARFKAIHFIHQPWYFTTTYNVVKPFLKSKLLERVFVHGDDLSGFYQEIDENILPSD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title