Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRALBP Antibody (OTI1G6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271275MFV450
Description
CRALBP Monoclonal antibody specifically detects CRALBP in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
CRALBP | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI1G6 | |
Flow Cytometry | |
Cellular retinaldehyde-binding protein, cellular retinaldehyde-binding protein-1, CRALBPretinaldehyde-binding protein 1, MGC3663, retinaldehyde binding protein 1 | |
Full length human recombinant protein of human RLBP1(NP_000317) produced in HEK293T cell. | |
0.1 mL | |
Vision | |
6017 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction