Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRALBP Polyclonal Antibody
CRALBP Polyclonal Antibody
Supplier: Thermo Scientific PA597184
Description
CRALBP Polyclonal Antibody for Western Blot

Specifications
CRALBP | |
Polyclonal | |
Unconjugated | |
RLBP1 | |
3110056M11Rik; cellular retinaldehyde-binding protein; cellular retinaldehyde-binding protein-1; CRALBP; retinaldehyde binding protein 1; retinaldehyde-binding protein 1; retinaldehyde-binding protein precursor; Rlbp1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19771, 293049 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Z275 | |
RLBP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human CRALBP (NP_0003171). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction