Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRAMP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559709 Supplier Invitrogen™ Supplier No. PA559709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRNPPRPLLV PGPSSTGSND SDGGLFAVPT TLPPNSRHGK LFSPSKEAEL TFRQHLNSIS MQSDFFLPKP RKLRNRHLRK PLVVQRTLLP RPSENQSHN Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

CRAMP1 (Cramped Chromatin Regulator Homolog 1) is a Protein Coding gene. Diseases associated with CRAMP1 include Deafness, Autosomal Dominant 40. An important paralog of this gene is ENSG00000261732. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRAMP1L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRAMP1
Gene Accession No. Q96RY5
Gene Alias C16orf34; CRAMP1; CRAMP1L; Cramped chromatin regulator homolog 1; Crm, cramped-like; hematological and neurological expressed 1-like protein; HN1L; KIAA1426; Protein cramped-like; TCE4; T-complex expressed gene 4
Gene Symbols CRAMP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRAMP1L. Recombinant protein control fragment (Product #RP-98610).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57585
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.