Learn More
CRAMP1L Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304093AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
This gene encodes a member of a highly conserved protein family that exists from archaea to vertebrates and plants. The encoded protein may function in RNA metabolism. Mutations within this gene are associated with Shwachman-Bodian-Diamond syndrome. An alternative transcript has been described, but its biological nature has not been determined. This gene has a closely linked pseudogene that is distally located.Specifications
CRAMP1L | |
Polyclonal | |
Unconjugated | |
CRAMP1 | |
CRAMP1L, Cramped chromatin regulator homolog 1, Crm (Cramped Drosophila)-like, Crm, cramped-like, Crm, cramped-like (Drosophila), hematological and neurological expressed 1-like protein, HN1L, KIAA1426, TCE4, T-complex expressed gene 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57585 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96RY5 | |
CRAMP1 | |
Between 300 and 350 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.