Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Craniofacial Development Protein 1 Antibody (11811), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243772
Description
Ensure accurate, reproducible results in Western Blot
Craniofacial Development Protein 1 Monoclonal specifically detects Craniofacial Development Protein 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Craniofacial Development Protein 1 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
CFDP1 | |
Recombinant protein encompassing a sequence within the center region of human CFDP1. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
11811 | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
BCNTBUCENTAUR, Bucentaur, CP27phosphoprotein (Bucentaur), craniofacial development protein 1, p97, SWC5, Yeti | |
Mouse | |
Protein G purified | |
RUO | |
10428 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction