Learn More
Crash Rescue Handgun Hydro-Chem™ Nozzles
Description
Pistol grip handline nozzles incorporate a foam solution or water application with simultaneous Hydro-Chem Technology – which projects dry chemical through the center of the foam/water stream – to achieve ranges not possible with conventional equipment.
Key Features
- Nozzles are made of hard, anodized aluminum alloys
- Feature 7 lb./sec dry chemical flow and a 1 in. NH (F) valved, dry chemical inlet; see ordering table for specifications related to foam solution flow and inlet
- Foam/water stream is adjustable from full fog to straight stream with shut off
Specifications
Specifications
| Flow Rate (English) | 95 gal./min. |
| Size | 1.5 in. |