Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRASP-1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990901
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0901 25 μL
50-199-0900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0901 Supplier Rockland Immunochemicals Supplier No. 200401C18S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for Borrelia burgdorferi CRASP-1 protein. A BLAST analysis was used to suggest reactivity with CRASP-1 from B. burgdorferi and B. garinii sources based on 100% homology with the immunizing sequence. Partial cross-reactivity is expected against B. spielmanii, afzelii, and valaisiana sources based on 98% homology. Cross-reactivity with CRASP-1 from other sources has not been determined.

CRASP-1, or Complement Regulator-Acquiring Surface Protein 1, is a multifunctional protein of Lyme disease-causing B. burgdorferi that binds to several human extracellular matrix proteins and plasminogen, including factor H (resulting in inhibition of complement activation in mammals) and Human Bone Morphogenic Protein 2. These interactions may contribute to adhesion, bacterial colonization, and organ tropism and may allow dissemination of B. burgdorferi in the host. B. burgdorferi spirochetes express up to 5 complement regulator-acquiring surface proteins. Multiple copies of sequences analagous to CRASP-1 genes have been detected in Borrelia plasmids. Borrelia species contain a large number of plasmids, of linear and circular, some of which appear to repeat sequences or contain fragments of other genes. These regions may serve as potentially usable information for the survival of Borrelia in its multiple environments during its life cycle. In addition, the sequence for CRASP-1 contains a repeated sequence folded into a stable stem loop structure typical of RNA genes.

Specifications

Antigen CRASP-1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen MBP-fusion protein corresponding to Borrelia burgdorferi CRASP-1 protein.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.