Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566557 Supplier Invitrogen™ Supplier No. PA566557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NHITLENISSG SSLNVKAGCV RKDWCESQPC QSRGRCINLW LSYQCDCHRP YEGPNCLREY VAGRFGQDDS TGYVIFTLDE SYGDTISLSM FVRTL Highest antigen sequence identity to the following orthologs - mouse 75%, rat 73%.

CRB1 (Crumbs homolog 1) plays a role in photoreceptor morphogenesis in the retina. It may maintain cell polarization and adhesion. CRB1 is similar to the Drosophila crumbs protein and localizes to the inner segment of mammalian photoreceptors. The first identified human homolog, CRB1, is expressed in retina and some parts of the brain, leaving room for another homolog to function in epithelial tissues. Mutations in this gene are associated with a severe form of retinitis pigmentosa, RP12, and with Leber congenital amaurosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRB1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRB1
Gene Accession No. P82279
Gene Alias CRB1; crumbs 1, cell polarity complex component; crumbs family member 1, photoreceptor morphogenesis associated; Crumbs homolog 1; LCA8; Protein crumbs homolog 1; RP12
Gene Symbols CRB1
Host Species Rabbit
Immunogen Recombinant Human CRB1. Recombinant protein control fragment (Product #RP-107215).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23418
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.