Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRB2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281710F
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
CRB2 Polyclonal antibody specifically detects CRB2 in Human, Mouse samples. It is validated for ELISASpecifications
CRB2 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
286204 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
crumbs homolog 2 (Drosophila) | |
Rabbit polyclonal CRB2 antibody was raised against a 17 amino acid peptide near the center of human CRB2. The immunogen is located within amino acids 460 - 510 of CRB2. Amino Acid Squence: AGTLATRNDTKESLELA | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction