Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525628
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525628 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525628 Supplier Invitrogen™ Supplier No. PA525628
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRB2 Polyclonal Antibody for IHC (P), Western Blot

CRB2 (Crumbs homolog 2), like its homologs CRB1 and CRB3, is similar to the Drosophila crumbs protein and is expressed in retina, brain and kidney. Along with other proteins, the Crumbs proteins form a complex that help set up cell polarity in developing neuroepithelial cells. At the onset of neural specification, embryonic stem cells (ESCs) upregulate CRB2, which then localizes apically in neural rosettes. Gain- and loss-of-function studies of CRB2 have shown that CRB2 is essential for the stabilization of other polarity proteins. Unlike CRB1, mutations in CRB2 do not appear to play a role in retinitis pigmentosa or in Leber congenital amaurosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CRB2
Gene Accession No. Q5IJ48, Q80YA8
Gene Alias 5930402A21; BC043114; Crb2; crumbs 2; crumbs 2, cell polarity complex component; crumbs family member 2; crumbs homolog 2; crumbs2; crumbs-like protein 2; FSGS9; Protein crumbs homolog 2; RGD1309368; VMCKD
Gene Symbols CRB2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1258-1285 amino acids of human CRUM2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241324, 286204
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.