Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA553092
Description
Immunogen sequence: VRKLREKRQT EGTYRPSSEE QFSHAAEARA PQDSKETVQG CLPI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
Involved in the establishment of cell polarity in mammalian epithelial cells. Regulates the morphogenesis of tight junctions.
Specifications
CRB3 | |
Polyclonal | |
Unconjugated | |
CRB3 | |
5730439B18; AU015179; AU041752; Crb3; crumbs 3, cell polarity complex component; crumbs family member 3; crumbs homolog 3; crumbs protein homolog 3; protein crumbs homolog 3; UNQ588/PRO1158 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92359 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUF7 | |
CRB3 | |
Recombinant protein corresponding to Human CRB3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction