Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CRBN Antibody Blocking Peptide
SDP

Catalog No. NB172147 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172147 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172147 Supplier Novus Biologicals™ Supplier No. NBP282079PEP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Blocking Peptied

Specifications

Accession Number NP_057386
Concentration 0.2 mg/ml
For Use With (Application) Block/Neutralize
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin
Gene ID (Entrez) 51185
Quantity 0.05 mg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias cereblon, DKFZp781K0715, MGC27358, MRT2A, non-syndromic, autosomal recessive, 2A, protein cereblon, protein x 0001
Gene Symbol CRBN
Research Category Neurodegeneration, Neuroscience
Protein CRBN
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.