Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRBN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561122 Supplier Invitrogen™ Supplier No. PA561122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVEDQDSKEA KKPNIINFDT SLPTSHTYLG ADMEEFHGRT LHDDDSCQVI PVLPQVMMIL IPGQTLPLQL FHPQEVSMVR NLIQKDRTFA VLAYSNVQER E Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

CRBN, a member of the Lon protease protein family, plays a role in brain development. It is a component of the DCX (DDB1-CUL4-X-box) E3 protein ligase complex, a complex that mediates the ubiquitination and subsequent proteasomal degradation of target proteins and is required for limb outgrowth and expression of the fibroblast growth factor FGF8. CRBN is thought to regulate the assembly and neuronal surface expression of large-conductance calcium-activated potassium channels in brain regions involved in memory and learning via its interaction with KCNT1. It is widely expressed and highly expressed in brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRBN
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRBN
Gene Accession No. Q56AP7, Q8C7D2, Q96SW2
Gene Alias 2610203G15Rik; 2900045O07Rik; AD-006; AF229032; AW108261; cereblon; CRBN; DKFZp781K0715; MGC27358; MRT2; MRT2A; novel lethal; piL; protein cereblon; Protein PiL; protein x 0001
Gene Symbols CRBN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRBN. Recombinant protein control fragment (Product #RP-96271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297498, 51185, 58799
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.