Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRBN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578492
Description
Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Mouse (92%), Rat (97%), Rhesus Monkey (99%), Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CRBN, a member of the Lon protease protein family, plays a role in brain development. It is a component of the DCX (DDB1-CUL4-X-box) E3 protein ligase complex, a complex that mediates the ubiquitination and subsequent proteasomal degradation of target proteins and is required for limb outgrowth and expression of the fibroblast growth factor FGF8. CRBN is thought to regulate the assembly and neuronal surface expression of large-conductance calcium-activated potassium channels in brain regions involved in memory and learning via its interaction with KCNT1. It is widely expressed and highly expressed in brain.
Specifications
CRBN | |
Polyclonal | |
Unconjugated | |
CRBN | |
2610203G15Rik; 2900045O07Rik; AD-006; AF229032; AW108261; cereblon; CRBN; DKFZp781K0715; MGC27358; MRT2; MRT2A; novel lethal; piL; protein cereblon; Protein PiL; protein x 0001 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297498, 51185, 58799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q56AP7, Q8C7D2, Q96SW2 | |
CRBN | |
Recombinant protein encompassing a sequence within the N-terminus region of human CRBN. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction