Learn More
CRBN Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 284941AP150UL
Description
The CRBN antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CRBN. This antibody recognizes human, mouse, rat antigen. The CRBN antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CRBN | |
Polyclonal | |
Unconjugated | |
150 μL | |
51185 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
333 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.